ADVH49B82DAB AChemBlock

(7-fluoroisoquinolin-6-yl)methanamine

  • Molecular Weight : 176.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-fluoroisoquinolin-6-yl-methanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72218
Availability details for
Quantity
  • Availability
  • Add to Cart