ADVH49B82DB3 AChemBlock

8-fluoroisoquinoline-6-carbaldehyde

  • Molecular Weight : 175.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-fluoroisoquinoline-6-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72234
Availability details for
Quantity
  • Availability
  • Add to Cart