ADVH49B82DB5 AChemBlock

6-(bromomethyl)-8-fluoroisoquinoline hydrobromide

  • Molecular Weight : 320.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromomethyl-8-fluoroisoquinoline-hydrobromide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72236
Availability details for
Quantity
  • Availability
  • Add to Cart