ADVH49B82DB8 AChemBlock

(8-fluoroisoquinolin-6-yl)boronic acid

  • Molecular Weight : 190.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-fluoroisoquinolin-6-yl-boronic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72249
Availability details for
Quantity
  • Availability
  • Add to Cart