ADVH49B82DB9 AChemBlock

8-(bromomethyl)-6-fluoroquinoline hydrobromide

  • Molecular Weight : 320.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-bromomethyl-6-fluoroquinoline-hydrobromide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72250
Availability details for
Quantity
  • Availability
  • Add to Cart