ADVH49B82DC0 AChemBlock

1-(4-(trifluoromethyl)isothiazol-3-yl)ethan-1-one

  • Molecular Weight : 195.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-trifluoromethyl-isothiazol-3-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72266
Availability details for
Quantity
  • Availability
  • Add to Cart