ADVH49B82DC1 AChemBlock

(4-(trifluoromethyl)isothiazol-3-yl)methanol

  • Molecular Weight : 183.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-trifluoromethyl-isothiazol-3-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72267
Availability details for
Quantity
  • Availability
  • Add to Cart