ADVH49B82DC3 AChemBlock

tert-butyl ((4-(trifluoromethyl)isothiazol-3-yl)methyl)carbamate

  • Molecular Weight : 282.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-trifluoromethyl-isothiazol-3-yl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72281
Availability details for
Quantity
  • Availability
  • Add to Cart