ADVH49B82DC7 AChemBlock

tert-butyl (R)-(3-bromo-6,7-dihydro-5H-pyrazolo[5,1-b][1,3]oxazin-6-yl)(methyl)carbamate

  • Molecular Weight : 332.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-r-3-bromo-6-7-dihydro-5h-pyrazolo-5-1-b-1-3-oxazin-6-yl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72293
Availability details for
Quantity
  • Availability
  • Add to Cart