ADVH49B82DC9 AChemBlock

Q 203 tosylate

  • Molecular Weight : 729.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/q-203-tosylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72390
Availability details for
Quantity
  • Availability
  • Add to Cart