ADVH49B82DCA AChemBlock

6-bromo-4-chloroindolin-2-one

  • Molecular Weight : 246.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-4-chloroindolin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72435
Availability details for
Quantity
  • Availability
  • Add to Cart