ADVH49B82DCF AChemBlock

1-Hexanesulfonamide

  • Molecular Weight : 165.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-hexanesulfonamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70409
Availability details for
Quantity
  • Availability
  • Add to Cart