ADVH49B82DD7 AChemBlock

1-Bromo-4-chloro-5-fluoro-2-trifluoromethyl-benzene

  • Molecular Weight : 277.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-bromo-4-chloro-5-fluoro-2-trifluoromethyl-benzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86648
Availability details for
Quantity
  • Availability
  • Add to Cart