ADVH49B82DEF AChemBlock

2-methoxy-4-(3-(trifluoromethyl)-3H-diazirin-3-yl)benzaldehyde

  • Molecular Weight : 244.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methoxy-4-3-trifluoromethyl-3h-diazirin-3-yl-benzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71908
Availability details for
Quantity
  • Availability
  • Add to Cart