ADVH49B82DF0 AChemBlock

2-amino-N-(2-((cyclopropylmethyl)(methyl)amino)ethyl)-2-methylpropanamide dihydrochloride

  • Molecular Weight : 286.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-n-2-cyclopropylmethyl-methyl-amino-ethyl-2-methylpropanamide-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86470
Availability details for
Quantity
  • Availability
  • Add to Cart