ADVH49B82E09 AChemBlock

2-amino-2-methyl-N-(2-(methyl(prop-2-yn-1-yl)amino)ethyl)propanamide dihydrochloride

  • Molecular Weight : 270.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-2-methyl-n-2-methyl-prop-2-yn-1-yl-amino-ethyl-propanamide-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86457
Availability details for
Quantity
  • Availability
  • Add to Cart