ADVH49B82E0F AChemBlock

2-{6-[4-(2-Amino-8-methoxy-quinazolin-4-yl)-[1,2,3]triazol-1-ylmethyl]-1-oxy-pyridin-2-yl}-propan-2-ol

  • Molecular Weight : 407.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-6-4-2-amino-8-methoxy-quinazolin-4-yl-1-2-3-triazol-1-ylmethyl-1-oxy-pyridin-2-yl-propan-2-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86658
Availability details for
Quantity
  • Availability
  • Add to Cart