ADVH49B82E18 AChemBlock

1-(2,4-Difluoro-benzyl)-cyclohexanecarboxylic acid

  • Molecular Weight : 254.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-4-difluoro-benzyl-cyclohexanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86620
Availability details for
Quantity
  • Availability
  • Add to Cart