ADVH49B82E19 AChemBlock

1-(2,4-Dichloro-phenyl)-4-[(pyrrolidine-1-sulfonylamino)-methyl]-5-[5-(4-trifluoromethyl-phenylethynyl)-thiophen-2-yl]-1H-pyrazole-3-carboxylic acid piperidin-1-ylamide

  • Molecular Weight : 751.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/1-2-4-dichloro-phenyl-4-pyrrolidine-1-sulfonylamino-methyl-5-5-4-trifluoromethyl-phenylethynyl-thiophen-2-yl-1h-pyrazole-3-carboxylic-acid-piperidin-1-ylamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86649
Availability details for
Quantity
  • Availability
  • Add to Cart