ADVH49B82E63 AChemBlock

(R)-tert-butyl 4-hydroxy-1-methoxybutan-2-ylcarbamate

  • Molecular Weight : 219.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-tert-butyl-4-hydroxy-1-methoxybutan-2-ylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72450
Availability details for
Quantity
  • Availability
  • Add to Cart