ADVH49B82E97 AChemBlock

3-(4-Methoxyphenyl)-2-thioxo-1,3-thiazolidin-4-one

  • Molecular Weight : 239.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-4-methoxyphenyl-2-thioxo-1-3-thiazolidin-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86868
Availability details for
Quantity
  • Availability
  • Add to Cart