ADVH49B82EAB AChemBlock

(4-Bromo-[1,5]naphthyridin-3-yl)-carbamic acid tert-butyl ester

  • Molecular Weight : 324.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1-5-naphthyridin-3-yl-carbamic-acid-tert-butyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86636
Availability details for
Quantity
  • Availability
  • Add to Cart