ADVH49B82EFC AChemBlock

(3-Fluoro-4-pyridin-2-yl-phenyl)-acetic acid

  • Molecular Weight : 231.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-4-pyridin-2-yl-phenyl-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86650
Availability details for
Quantity
  • Availability
  • Add to Cart