ADVH49B82F4A AChemBlock

(2R)-2-amino-2-(1-methyl-1H-pyrazol-4-yl)acetic acid hydrochloride

  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-2-amino-2-1-methyl-1h-pyrazol-4-yl-acetic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86102
Availability details for
Quantity
  • Availability
  • Add to Cart