ADVH49B82F4B AChemBlock

(2R)-2-{[(benzyloxy)carbonyl]amino}-3-{1-[(tert-butoxy)carbonyl]piperidin-4-yl}propanoic acid

  • Molecular Weight : 406.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-2-benzyloxy-carbonyl-amino-3-1-tert-butoxy-carbonyl-piperidin-4-yl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86104
Availability details for
Quantity
  • Availability
  • Add to Cart