ADVH49B82F4C AChemBlock

(2R)-2-[[(1S,2S)-2-(dipropylamino)cyclohexyl]carbamothioylamino]-N,3,3-trimethyl-butanamide

  • Molecular Weight : 384.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-2-1s-2s-2-dipropylamino-cyclohexyl-carbamothioylamino-n-3-3-trimethyl-butanamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86105
Availability details for
Quantity
  • Availability
  • Add to Cart