ADVH49B82F4D AChemBlock

(2R)-2-[(2R)-pyrrolidin-2-yl]pyrrolidine dihydrochloride

  • Molecular Weight : 213.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-2-2r-pyrrolidin-2-yl-pyrrolidine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86106
Availability details for
Quantity
  • Availability
  • Add to Cart