ADVH49B82F51 AChemBlock

(2R)-2-(bromomethyl)oxetane

  • Molecular Weight : 151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-2-bromomethyl-oxetane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86109
Availability details for
Quantity
  • Availability
  • Add to Cart