ADVH49B82F53 AChemBlock

(2R)-1-[(benzyloxy)carbonyl]-5-oxopiperidine-2-carboxylic acid

  • Molecular Weight : 277.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-1-benzyloxy-carbonyl-5-oxopiperidine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86111
Availability details for
Quantity
  • Availability
  • Add to Cart