ADVH49B82F54 AChemBlock

(2R)-1-[(benzyloxy)carbonyl]-2-methylpiperidine-4-carboxylic acid

  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-1-benzyloxy-carbonyl-2-methylpiperidine-4-carboxylic-acid-309422.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86112
Availability details for
Quantity
  • Availability
  • Add to Cart