ADVH49B82F57 AChemBlock

(2E)-3-[(2R)-1-methylpyrrolidin-2-yl]prop-2-enoic acid hydrochloride

  • Molecular Weight : 191.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2e-3-2r-1-methylpyrrolidin-2-yl-prop-2-enoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86115
Availability details for
Quantity
  • Availability
  • Add to Cart