ADVH49B82F59 AChemBlock

(2-chloro-3-oxo-cyclohexen-1-yl) trifluoromethanesulfonate

  • Molecular Weight : 278.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-3-oxo-cyclohexen-1-yl-trifluoromethanesulfonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86117
Availability details for
Quantity
  • Availability
  • Add to Cart