ADVH49B82F5A AChemBlock

(2-bromo-3,5-difluoro-4-pyridyl)methanol

  • Molecular Weight : 224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-3-5-difluoro-4-pyridyl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86118
Availability details for
Quantity
  • Availability
  • Add to Cart