ADVH49B82F64 AChemBlock

(1S,3S)-rel-3-amino-1-methylcyclopentane-1-carboxylic acid

  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-3s-rel-3-amino-1-methylcyclopentane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86128
Availability details for
Quantity
  • Availability
  • Add to Cart