ADVH49B82F66 AChemBlock

(1S,3S)-rel-3-(tert-butoxycarbonylamino)-1-methyl-cyclohexanecarboxylic acid

  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-3s-rel-3-tert-butoxycarbonylamino-1-methyl-cyclohexanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86130
Availability details for
Quantity
  • Availability
  • Add to Cart