ADVH49B82F68 AChemBlock

(1s,3s)-3-Amino-cyclohexanecarboxylic acid

  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1s-3s-3-amino-cyclohexanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86621
Availability details for
Quantity
  • Availability
  • Add to Cart