ADVH49B82F6A AChemBlock

(1S,3R)-3-amino-N,1-dimethyl-cyclopentanecarboxamide hydrochloride

  • Molecular Weight : 192.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-3r-3-amino-n-1-dimethyl-cyclopentanecarboxamide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86133
Availability details for
Quantity
  • Availability
  • Add to Cart