ADVH49B82F6E AChemBlock

2-Chloro-5-fluoro-quinazolin-4-ol

  • Molecular Weight : 198.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-fluoro-quinazolin-4-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86622
Availability details for
Quantity
  • Availability
  • Add to Cart