ADVH49B82F82 AChemBlock

(1S)-1-(pyrimidin-5-yl)ethan-1-amine hydrochloride

  • Molecular Weight : 159.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-1-pyrimidin-5-yl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86159
Availability details for
Quantity
  • Availability
  • Add to Cart