ADVH49B82F84 AChemBlock

(1R,8aR)-2-[(tert-butoxy)carbonyl]-octahydropyrrolo[1,2-a]piperazine-1-carboxylic acid

  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-8ar-2-tert-butoxy-carbonyl-octahydropyrrolo-1-2-a-piperazine-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86161
Availability details for
Quantity
  • Availability
  • Add to Cart