ADVH49B82F89 AChemBlock

(1R,4R)-5,5-difluoro-2-azabicyclo[2.2.1]heptane hydrochloride

  • Molecular Weight : 169.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-4r-5-5-difluoro-2-azabicyclo-2-2-1-heptane-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86170
Availability details for
Quantity
  • Availability
  • Add to Cart