ADVH49B82F8A AChemBlock

(1R,3s,5S)-rel-9-[(tert-butoxy)carbonyl]-9-azabicyclo[3.3.1]nonane-3-carboxylic acid

  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-3s-5s-rel-9-tert-butoxy-carbonyl-9-azabicyclo-3-3-1-nonane-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86172
Availability details for
Quantity
  • Availability
  • Add to Cart