ADVH49B82F8C AChemBlock

(1R,3S,4R)-3-(tert-butoxycarbonylamino)-4-hydroxy-cyclopentanecarboxylic acid 2-methylpropan-2-amine

  • Molecular Weight : 318.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-3s-4r-3-tert-butoxycarbonylamino-4-hydroxy-cyclopentanecarboxylic-acid-2-methylpropan-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86174
Availability details for
Quantity
  • Availability
  • Add to Cart