ADVH49B82F8E AChemBlock

(1R,3r,5S)-rel-6-(benzyloxy)bicyclo[3.1.0]hexan-3-ol

  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-5s-rel-6-benzyloxy-bicyclo-3-1-0-hexan-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86176
Availability details for
Quantity
  • Availability
  • Add to Cart