ADVH49B82F91 AChemBlock

(1R,3R)-cyclohexane-1,3-dicarboxylic acid

  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-cyclohexane-1-3-dicarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86179
Availability details for
Quantity
  • Availability
  • Add to Cart