ADVH49B82F94 AChemBlock

(1R,3R)-3-aminocyclohexane-1-carboxylic acid hydrochloride

  • Molecular Weight : 179.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-3-aminocyclohexane-1-carboxylic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86182
Availability details for
Quantity
  • Availability
  • Add to Cart