ADVH49B82F95 AChemBlock

(1R,3R)-3-amino-1-(trifluoromethyl)cyclohexanol

  • Molecular Weight : 183.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-3-amino-1-trifluoromethyl-cyclohexanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86183
Availability details for
Quantity
  • Availability
  • Add to Cart