ADVH49B82F96 AChemBlock

(1r,2s,5s)-rel-3-boc-3-azabicyclo[3.1.0]hexane-2-methanol

  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-2s-5s-rel-3-boc-3-azabicyclo-3-1-0-hexane-2-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86184
Availability details for
Quantity
  • Availability
  • Add to Cart