ADVH49B82F98 AChemBlock

(1R,2S,5R)-rel-8-methyl-8-azabicyclo[3.2.1]octan-2-amine dihydrochloride

  • Molecular Weight : 213.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-2s-5r-rel-8-methyl-8-azabicyclo-3-2-1-octan-2-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86186
Availability details for
Quantity
  • Availability
  • Add to Cart