ADVH49B82F9B AChemBlock

(1R,2R)-ethyl 2-(tert-butoxycarbonyl)cyclohexanecarboxylate

  • Molecular Weight : 256.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1r-2r-ethyl-2-tert-butoxycarbonyl-cyclohexanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72452
Availability details for
Quantity
  • Availability
  • Add to Cart